1-(3-Methoxythien-2-yl)ethanone

Lieferant: Apollo Scientific

OR110057-1G
APOSOR110057-1GEA 223 EUR
APOSOR110057-1G
1-(3-Methoxythien-2-yl)ethanone
1-(3-Methoxythien-2-yl)ethanone
Formel: C₇H₈O₂S
Molecular Weight: 156,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16658745
CAS-Nummer: 51514-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR