7-Chlorimidazo[1,2-a]pyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR110058-25G
APOSOR110058-25GEA
122
EUR
APOSOR110058-25G
7-Chlorimidazo[1,2-a]pyridin
7-Chlorimidazo[1,2-a]pyridin
Formel:
C₇H₅ClN₂ Schmelzpunkt: 49…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08275126 CAS-Nummer: 4532-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...