4-{[4-(Hydroxymethyl)phenyl]amino}-4-oxobut-2-enoic acid
Lieferant: Apollo Scientific
OR110062-1G
APOSOR110062-1GEA
232
EUR
APOSOR110062-1G
4-{[4-(Hydroxymethyl)phenyl]amino}-4-oxobut-2-enoic acid
4-{[4-(Hydroxymethyl)phenyl]amino}-4-oxobut-2-enoic acid
Formel:
C₁₁H₁₁NO₄ Molecular Weight: 221,209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24850051 CAS-Nummer: 1440537-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...