5-Bromo-1H-indole-2-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
OR110064-1G
APOSOR110064-1GEA
464
EUR
APOSOR110064-1G
5-Bromo-1H-indole-2-carbohydrazide
5-Bromo-1H-indole-2-carbohydrazide
Formel:
C₉H₈BrN₃O Molecular Weight: 254,083 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07364659 CAS-Nummer: 20948-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...