Ethyl 5-bromo-2-ethoxynicotinate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 5-bromo-2-ethoxy-3-pyridinecarboxylate
OR110066-1G
APOSOR110066-1GEA
346
EUR
APOSOR110066-1G
Ethyl 5-bromo-2-ethoxynicotinate
Ethyl 5-bromo-2-ethoxynicotinate
Formel:
C₁₀H₁₂BrNO₃ Molecular Weight: 274,111 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19105409 CAS-Nummer: 1310416-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...