4-(4-Bromo-3-methylphenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Lieferant: Apollo Scientific
OR110067-1G
APOSOR110067-1GEA
232
EUR
APOSOR110067-1G
4-(4-Bromo-3-methylphenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
4-(4-Bromo-3-methylphenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Formel:
C₉H₈BrN₃O Molecular Weight: 254,083 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23381089 CAS-Nummer: 1440535-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...