4-{[3-(Methoxycarbonyl)thien-2-yl]amino}-4-oxobut-2-enoic acid

Lieferant: Apollo Scientific

OR110068-1G
APOSOR110068-1GEA 241 EUR
APOSOR110068-1G
4-{[3-(Methoxycarbonyl)thien-2-yl]amino}-4-oxobut-2-enoic acid
4-{[3-(Methoxycarbonyl)thien-2-yl]amino}-4-oxobut-2-enoic acid
Formel: C₁₀H₉NO₅S
Molecular Weight: 255,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07801526
CAS-Nummer: 590376-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR