2-Morpholin-4-yl-5-(morpholin-4-ylsulphonyl)benzoic acid

Lieferant: Apollo Scientific

OR110070-1G
APOSOR110070-1GEA 248 EUR
APOSOR110070-1G
2-Morpholin-4-yl-5-(morpholin-4-ylsulphonyl)benzoic acid
2-Morpholin-4-yl-5-(morpholin-4-ylsulphonyl)benzoic acid
Formel: C₁₅H₂₀N₂O₆S
Molecular Weight: 356.394 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06372770
CAS-Nummer: 796106-55-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR