3-Brom-7-chlorimidazo[1,2-a]pyridin

Lieferant: Apollo Scientific
Achtung

OR110071-1G
APOSOR110071-1GEA 168 EUR
APOSOR110071-1G
3-Brom-7-chlorimidazo[1,2-a]pyridin
3-Brom-7-chlorimidazo[1,2-a]pyridin
Formel: C₇H₄BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11111834
CAS-Nummer: 342613-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR