4-[(1-Methyl-1H-indazol-3-yl)amino]-4-oxobutanoic acid

Lieferant: Apollo Scientific

OR110072-1G
APOSOR110072-1GEA 0 EUR
APOSOR110072-1G
4-[(1-Methyl-1H-indazol-3-yl)amino]-4-oxobutanoic acid
4-[(1-Methyl-1H-indazol-3-yl)amino]-4-oxobutanoic acid
Formel: C₁₂H₁₃N₃O₃
Molecular Weight: 247,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24850052
CAS-Nummer: 1440535-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR