Tetrahydropyran-2-ylmethylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR110077-1G
APOSOR110077-1GEA 296 EUR
APOSOR110077-1G
Tetrahydropyran-2-ylmethylamine hydrochloride
Tetrahydropyran-2-ylmethylamine hydrochloride
Formel: C₆H₁₃NO·ClH
Schmelzpunkt: 142…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06738971
CAS-Nummer: 683233-12-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR