2-(Tetrahydro-2H-pyran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione

Lieferant: Apollo Scientific

OR110078-1G
APOSOR110078-1GEA 296 EUR
APOSOR110078-1G
2-(Tetrahydro-2H-pyran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione
2-(Tetrahydro-2H-pyran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione
Formel: C₁₄H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11277601
CAS-Nummer: 111293-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR