2-Thien-2-ylimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

OR110079-1G
APOSOR110079-1GEA 296 EUR
APOSOR110079-1G
2-Thien-2-ylimidazo[1,2-a]pyridine
2-Thien-2-ylimidazo[1,2-a]pyridine
Formel: C₁₁H₈N₂S
Molecular Weight: 200,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00507037
CAS-Nummer: 4045-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR