3-Cyanphenylboronsäurepinakolester 97%

Lieferant: Apollo Scientific

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 3-Cyanobenzeneboronic acid pinacol ester

OR11008-25G
APOSOR11008-25GEA 56.7 EUR
APOSOR11008-25G
3-Cyanphenylboronsäurepinakolester 97%
3-Cyanphenylboronsäurepinakolester
Formel: C₁₃H₁₆BNO₂
Molecular Weight: 229,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03789262
CAS-Nummer: 214360-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR