3,5,6,8-Tetrahydro-4H-pyrano[4',3':4,5]thieno[2,3-d]pyrimidin-4-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Oxo-3,5,6,8-tetrahydro-4H-pyrano[4',3':4,5]thieno[2,3-d]pyrimidine

OR110080-1G
APOSOR110080-1GEA 1120 EUR
APOSOR110080-1G
3,5,6,8-Tetrahydro-4H-pyrano[4',3':4,5]thieno[2,3-d]pyrimidin-4-one
3,5,6,8-Tetrahydro-4H-pyrano[4',3':4,5]thieno[2,3-d]pyrimidin-4-one
Formel: C₉H₈N₂O₂S
Molecular Weight: 208,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16251081
CAS-Nummer: 243968-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR