1-Acetyl-5-(morpholin-4-ylsulphonyl)indoline

Lieferant: Apollo Scientific

OR110081-1G
APOSOR110081-1GEA 296 EUR
APOSOR110081-1G
1-Acetyl-5-(morpholin-4-ylsulphonyl)indoline
1-Acetyl-5-(morpholin-4-ylsulphonyl)indoline
Formel: C₁₄H₁₈N₂O₄S
Molecular Weight: 310.369 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05857252
CAS-Nummer: 698983-77-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR