2-Chloro-N-(1-methyl-1H-indazol-5-yl)acetamide
Lieferant: Apollo Scientific
OR110084-1G
APOSOR110084-1GEA
290
EUR
APOSOR110084-1G
2-Chloro-N-(1-methyl-1H-indazol-5-yl)acetamide
2-Chloro-N-(1-methyl-1H-indazol-5-yl)acetamide
Formel:
C₁₀H₁₀ClN₃O Molecular Weight: 223,659 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22577290 CAS-Nummer: 1427460-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...