2-Methyl-1,3-benzothiazole-6-sulphonamide

Lieferant: Apollo Scientific

OR110086-1G
APOSOR110086-1GEA 290 EUR
APOSOR110086-1G
2-Methyl-1,3-benzothiazole-6-sulphonamide
2-Methyl-1,3-benzothiazole-6-sulphonamide
Formel: C₈H₈N₂O₂S₂
Molecular Weight: 228,291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381086
CAS-Nummer: 21431-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR