1-tert-Butoxycarbonyl-4-{[(tetrahydrofuran-2-ylmethyl)amino]methyl}piperidin-4-ol

Lieferant: Apollo Scientific

OR110087-1G
APOSOR110087-1GEA 310 EUR
APOSOR110087-1G
1-tert-Butoxycarbonyl-4-{[(tetrahydrofuran-2-ylmethyl)amino]methyl}piperidin-4-ol
1-tert-Butoxycarbonyl-4-{[(tetrahydrofuran-2-ylmethyl)amino]methyl}piperidin-4-ol
Formel: C₁₆H₃₀N₂O₄
Molecular Weight: 314,42 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381092
CAS-Nummer: 1440535-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR