tert-Butyl 3-[4-(ethoxycarbonyl)piperidin-1-yl]pyrrolidine-1-carboxylate
Lieferant: Apollo Scientific
OR110088-1G
APOSOR110088-1GEA
310
EUR
APOSOR110088-1G
tert-Butyl 3-[4-(ethoxycarbonyl)piperidin-1-yl]pyrrolidine-1-carboxylate
tert-Butyl 3-[4-(ethoxycarbonyl)piperidin-1-yl]pyrrolidine-1-carboxylate
Formel:
C₁₇H₃₀N₂O₄ Molecular Weight: 326,431 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24850056 CAS-Nummer: 1440535-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...