Methyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110090-5G
APOSOR110090-5GEA 0 EUR
APOSOR110090-5G
Methyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate
Methyl 7-bromoimidazo[1,2-a]pyridine-3-carboxylate
Formel: C₉H₇BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20527654
CAS-Nummer: 1313410-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR