Ethyl 8-bromoimidazo[1,2-a]pyridine-6-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110093-1G
APOSOR110093-1GEA 317 EUR
APOSOR110093-1G
Ethyl 8-bromoimidazo[1,2-a]pyridine-6-carboxylate
Ethyl 8-bromoimidazo[1,2-a]pyridine-6-carboxylate
Formel: C₁₀H₉BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23159159
CAS-Nummer: 957103-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR