Ethyl-3-amino-4-iodo-1-benzothiophene-2-carboxylate
Lieferant: Apollo Scientific
OR110095-1G
APOSOR110095-1GEA
275
EUR
APOSOR110095-1G
Ethyl-3-amino-4-iodo-1-benzothiophene-2-carboxylate
Ethyl-3-amino-4-iodo-1-benzothiophene-2-carboxylate
Formel:
C₁₁H₁₀INO₂S Molecular Weight: 347,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505027 CAS-Nummer: 1216596-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...