Methyl-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate

OR110097-250MG
APOSOR110097-250MGEA 34.8 EUR
APOSOR110097-250MG
Methyl-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate
Methyl-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylate
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,2000 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08544342
CAS-Nummer: 758684-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR