Furan-3-boronsäurepinakolester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(3-Furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan
OR11010-25G
APOSOR11010-25GEA
217
EUR
APOSOR11010-25G
Furan-3-boronsäurepinakolester
Furan-3-boronsäurepinakolester
Formel:
C₁₀H₁₅BO₃ Schmelzpunkt: 62…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03453669 CAS-Nummer: 248924-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...