Methyl 2-acetyl-5,7-dibromo-2H-indazole-6-carboxylate

Lieferant: Apollo Scientific

OR110100-1G
APOSOR110100-1GEA 0 EUR
APOSOR110100-1G
Methyl 2-acetyl-5,7-dibromo-2H-indazole-6-carboxylate
Methyl 2-acetyl-5,7-dibromo-2H-indazole-6-carboxylate
Formel: C₁₁H₈Br₂N₂O₃
Molecular Weight: 376,001 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23159163
CAS-Nummer: 1427460-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR