Ethyl 3-bromo-7-chloroimidazo[1,2-a]pyridine-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR110101-1G
APOSOR110101-1GEA
348
EUR
APOSOR110101-1G
Ethyl 3-bromo-7-chloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-bromo-7-chloroimidazo[1,2-a]pyridine-2-carboxylate
Formel:
C₁₀H₈BrClN₂O₂ Molecular Weight: 303,54 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23159165 CAS-Nummer: 1427460-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...