1-(1-Methyl-1H-indazol-3-yl)-1H-pyrrole-2,5-dione

Lieferant: Apollo Scientific

OR110108-1G
APOSOR110108-1GEA 348 EUR
APOSOR110108-1G
1-(1-Methyl-1H-indazol-3-yl)-1H-pyrrole-2,5-dione
1-(1-Methyl-1H-indazol-3-yl)-1H-pyrrole-2,5-dione
Formel: C₁₂H₉N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD24850057
CAS-Nummer: 1440535-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR