5-Bromo-1-methyl-1H-indole-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110109-250MG
APOSOR110109-250MGEA 232 EUR
APOSOR110109-250MG
5-Bromo-1-methyl-1H-indole-2-carbohydrazide
5-Bromo-1-methyl-1H-indole-2-carbohydrazide
Formel: C₁₀H₁₀BrN₃O
Molecular Weight: 268,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24850058
CAS-Nummer: 1440535-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR