2-Methyl-4-nitro-2H-indazole
Lieferant: Apollo Scientific
OR110110-1G
APOSOR110110-1GEA
217
EUR
APOSOR110110-1G
2-Methyl-4-nitro-2H-indazole
2-Methyl-4-nitro-2H-indazole
Formel:
C₈H₇N₃O₂ Molecular Weight: 177,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11982555 CAS-Nummer: 26120-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...