Methyl 7-chloro-1H-indazole-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR110122-250MG
APOSOR110122-250MGEA
265
EUR
APOSOR110122-250MG
Methyl 7-chloro-1H-indazole-5-carboxylate
Methyl 7-chloro-1H-indazole-5-carboxylate
Formel:
C₉H₇ClN₂O₂ Molecular Weight: 210,617 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11976871 CAS-Nummer: 1260851-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...