1-Methyl-1H-indole-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110130-100MG
APOSOR110130-100MGEA 325 EUR
APOSOR110130-100MG
1-Methyl-1H-indole-2-carbohydrazide
1-Methyl-1H-indole-2-carbohydrazide
Formel: C₁₀H₁₁N₃O
Molecular Weight: 189.214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463397
CAS-Nummer: 56809-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR