2-Butyl-6-nitrobenzoxazol

Lieferant: Apollo Scientific

OR110139-10G OR110139-1G OR110139-5G
APOSOR110139-10GEA 0 EUR
APOSOR110139-10G APOSOR110139-1G APOSOR110139-5G
2-Butyl-6-nitrobenzoxazol
2-Butyl-6-nitrobenzoxazole
Formel: C₁₁H₁₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05663809
CAS-Nummer: 886360-96-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR