4-Chloro-6-[4-(2-hydroxyethyl)piperazin-1-yl]pyrimidine-5-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR110141-1G
APOSOR110141-1GEA 0 EUR
APOSOR110141-1G
4-Chloro-6-[4-(2-hydroxyethyl)piperazin-1-yl]pyrimidine-5-carbaldehyde
4-Chloro-6-[4-(2-hydroxyethyl)piperazin-1-yl]pyrimidine-5-carbaldehyde
Formel: C₁₁H₁₅ClN₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18074535
CAS-Nummer: 1427460-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR