1-Methyl-5-nitro-1H-indazol-3-amine

Lieferant: Apollo Scientific

OR110142-1G
APOSOR110142-1GEA 91.5 EUR
APOSOR110142-1G
1-Methyl-5-nitro-1H-indazol-3-amine
1-Methyl-5-nitro-1H-indazol-3-amine
Formel: C₈H₈N₄O₂
Molecular Weight: 192,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619579
CAS-Nummer: 73105-48-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR