4-Methyl-2-phenyl-1,3-oxazole
Lieferant: Apollo Scientific
|
Achtung
|
OR110144-1G
APOSOR110144-1GEA
0
EUR
APOSOR110144-1G
4-Methyl-2-phenyl-1,3-oxazole
4-Methyl-2-phenyl-1,3-oxazole
Formel:
C₁₀H₉NO Siedepunkt: 60 °C (1 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD11055268 CAS-Nummer: 877-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...