4-(1-Pyrrolyl)benzonitril

Lieferant: Apollo Scientific

OR110146-10G OR110146-1G OR110146-5G
APOSOR110146-10GEA 0 EUR
APOSOR110146-10G APOSOR110146-1G APOSOR110146-5G
4-(1-Pyrrolyl)benzonitril
4-(1-Pyrrolyl)benzonitril
Formel: C₁₁H₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00085164
CAS-Nummer: 23351-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR