Methyl-8-bromimidazo[1,2-a]pyridin-6-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 8-bromoimidazo[1,2-a]pyridine-6-carboxylate

OR110150-1G
APOSOR110150-1GEA 109 EUR
APOSOR110150-1G
Methyl-8-bromimidazo[1,2-a]pyridin-6-carboxylat
Methyl-8-bromimidazo[1,2-a]pyridin-6-carboxylat
Formel: C₉H₇BrN₂O₂
Molecular Weight: 255,0681 g/mol
Schmelzpunkt: 125…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD17016026
CAS-Nummer: 1234616-08-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR