3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carboxylic acid
OR110152-1G
APOSOR110152-1GEA
68.3
EUR
APOSOR110152-1G
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carboxylic acid
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carboxylic acid
Formel:
C₉H₇NO₄ Molecular Weight: 193,156 g/mol Schmelzpunkt: 301…304 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03036647 CAS-Nummer: 134997-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...