[(7-Chloroquinolin-4-yl)thio]acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR110153-1G
APOSOR110153-1GEA
65.4
EUR
APOSOR110153-1G
[(7-Chloroquinolin-4-yl)thio]acetic acid
[(7-Chloroquinolin-4-yl)thio]acetic acid
Formel:
C₁₁H₈ClNO₂S Molecular Weight: 253,705 g/mol Schmelzpunkt: 223…225 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569501 CAS-Nummer: 5429-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...