(2E)-1-(4-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one
Lieferant: Apollo Scientific
Synonyme:
(E)-3-(4-Chlorophenyl)-N,N-dimethylacrylamide, (E)-1-(4-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one, (E)-1-(4-Chlorophenyl)-3-dimethylaminopropenone, 1-(4-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one
OR110156-1G
OR110156-5G
OR110156-10G
APOSOR110156-1GEA
63.8
EUR
APOSOR110156-1G
APOSOR110156-5G
APOSOR110156-10G
(2E)-1-(4-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one
(2E)-1-(4-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one
Formel:
C₁₁H₁₂ClNO Molecular Weight: 209.68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00172116 CAS-Nummer: 28587-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...