3-Amino-3-(3-bromphenyl)propionsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: DL-3-Amino-3-(3-Bromophenyl)propionic acid

OR110160-1G
APOSOR110160-1GEA 21.9 EUR
APOSOR110160-1G
3-Amino-3-(3-bromphenyl)propionsäure
3-Amino-3-(3-bromphenyl)propionsäure
Formel: C₉H₁₀BrNO₂
Molecular Weight: 244,0852 g/mol
Schmelzpunkt: 218…220 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00995843
CAS-Nummer: 117391-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR