Ethyl 3-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110161-1G
APOSOR110161-1GEA 360 EUR
APOSOR110161-1G
Ethyl 3-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate
Formel: C₁₁H₁₁BrN₂O₂
Molecular Weight: 283,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20526410
CAS-Nummer: 1123167-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR