5-Methylimidazo[1,2-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific

OR110162-1G
APOSOR110162-1GEA 75.4 EUR
APOSOR110162-1G
5-Methylimidazo[1,2-a]pyridine-2-carboxylic acid
5-Methylimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C9H8N2O2
Molecular Weight: 176,1720 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419472
CAS-Nummer: 88751-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR