Ethyl 6,8-dibromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110166-1G
APOSOR110166-1GEA 140 EUR
APOSOR110166-1G
Ethyl 6,8-dibromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6,8-dibromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate
Formel: C₁₁H₁₀Br₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22421937
CAS-Nummer: 859787-43-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR