Ethyl 3-(2-oxoimidazolidin-1-yl)benzoate

Lieferant: Apollo Scientific
Achtung

OR110168-1G
APOSOR110168-1GEA 476 EUR
APOSOR110168-1G
Ethyl 3-(2-oxoimidazolidin-1-yl)benzoate
Ethyl 3-(2-oxoimidazolidin-1-yl)benzoate
Formel: C₁₂H₁₄N₂O₃
Molecular Weight: 234,251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22421940
CAS-Nummer: 1427460-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR