2-Methyl-1,3-benzoxazole-5-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR110177-1G
APOSOR110177-1GEA
290
EUR
APOSOR110177-1G
2-Methyl-1,3-benzoxazole-5-carbonitrile
2-Methyl-1,3-benzoxazole-5-carbonitrile
Formel:
C₉H₆N₂O Schmelzpunkt: 115…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11035941 CAS-Nummer: 903556-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...