2-Methyloxazolo[4,5-b]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR110178-1G
APOSOR110178-1GEA
186
EUR
APOSOR110178-1G
2-Methyloxazolo[4,5-b]pyridine
2-Methyloxazolo[4,5-b]pyridine
Formel:
C₇H₆N₂O Schmelzpunkt: 65…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05256196 CAS-Nummer: 86467-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...