2-Ethyl[1,3]oxazolo[4,5-b]pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 2-Ethyloxazolo[4,5-b]pyridine

OR110179-5G
APOSOR110179-5GEA 108 EUR
APOSOR110179-5G
2-Ethyl[1,3]oxazolo[4,5-b]pyridine 95%
2-Ethyl[1,3]oxazolo[4,5-b]pyridine
Formel: C₈H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11110208
CAS-Nummer: 52333-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR