1,2,3,4-Tetrahydroisoquinolin-5-amine

Lieferant: Apollo Scientific

Synonyme: 5-Amino-1,2,3,4-tetrahydroisoquinoline

OR110180-1G
APOSOR110180-1GEA 139 EUR
APOSOR110180-1G
1,2,3,4-Tetrahydroisoquinolin-5-amine
1,2,3,4-Tetrahydroisoquinolin-5-amine
Formel: C₉H₁₂N₂
Molecular Weight: 148.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684185
CAS-Nummer: 115955-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR